The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145970.1 | NP_001139442.1 | 1 - 2 | 305 | 94 |
NM_144941.3 | NP_659190.3 | 1 - 2 | 305 | 94 | |
GenBank mRNA | AK151613.1 | - | 1 - 2 | 293 | 94 |
BC023677.1 | - | 1 - 2 | 280 | 94 | |
BC023774.1 | - | 1 - 2 | 302 | 94 | |
BU053928.1 | - | 1 - 2 | 305 | 94 |