The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026601.3 | NP_080877.3 | - | 342 | 65 |
GenBank mRNA | AA403499.1 | - | 2 - 3 | 351 | 65 |
BC024619.1 | - | - | 330 | 65 | |
BC100401.1 | - | - | 306 | 65 |