The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030260.3 | NP_084536.2 | 5 - 6 | 1601 | 124 |
NM_173002.3 | NP_766590.1 | 5 - 6 | 1601 | 124 | |
GenBank mRNA | AK030287.1 | - | 5 - 6 | 1517 | 124 |
AK042190.1 | - | 5 - 6 | 1528 | 124 | |
AK042392.1 | - | 5 - 6 | 1511 | 124 | |
AK044429.1 | - | 5 - 6 | 1494 | 124 | |
AK044529.1 | - | 5 - 6 | 1525 | 124 | |
AK044948.1 | - | 5 - 6 | 1495 | 124 | |
AK047079.1 | - | 5 - 6 | 1483 | 124 | |
AK047538.1 | - | 5 - 6 | 1517 | 124 | |
AK080307.1 | - | 5 - 6 | 1495 | 124 | |
AK081005.1 | - | 5 - 6 | 1603 | 124 | |
AK169517.1 | - | 5 - 6 | 1488 | 124 | |
BC003332.1 | - | 5 - 6 | 1585 | 124 |