Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec4a2
Entrez Gene ID
26888
Gene Name
C-type lectin domain family 4, member a2
Gene Aliases
Clec4a, Clecsf6, DCIR, Dcir1
Chromosome Location
Chr.6: 123122690 - 123143999 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170332.1 NP_001163803.1 1 - 2 64 147
GenBank mRNA BC096565.1 - 1 - 2 73 147

View Full Product Details