Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc24
Entrez Gene ID
381546
Gene Name
coiled-coil domain containing 24
Gene Aliases
Gm1028
Chromosome Location
Chr.4: 117869260 - 117872661 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034876.1 NP_001030048.1 5 - 6 564 62
GenBank mRNA AK133118.1 - 5 - 6 565 62
BC026478.1 - - 288 62

View Full Product Details