The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253681.1 | NP_001240610.1 | 3 - 4 | 459 | 94 |
NM_001253686.1 | NP_001240615.1 | 3 - 4 | 459 | 94 | |
NM_001253687.1 | NP_001240616.1 | 3 - 4 | 459 | 94 | |
NM_001253688.1 | NP_001240617.1 | 3 - 4 | 459 | 94 | |
NM_001253689.1 | NP_001240618.1 | 3 - 4 | 459 | 94 | |
NM_029673.3 | NP_083949.2 | 3 - 4 | 459 | 94 | |
GenBank mRNA | AK030841.1 | - | 3 - 4 | 401 | 94 |
AK033126.1 | - | 3 - 4 | 370 | 94 | |
AK038129.1 | - | 3 - 4 | 449 | 94 | |
AK049189.1 | - | 3 - 4 | 403 | 94 | |
AK146640.1 | - | 3 - 4 | 451 | 94 | |
AK152588.1 | - | 3 - 4 | 405 | 94 | |
AK157805.1 | - | 3 - 4 | 400 | 94 | |
AK159909.1 | - | 3 - 4 | 410 | 94 | |
AK163491.1 | - | 3 - 4 | 367 | 94 | |
AK169365.1 | - | 3 - 4 | 362 | 94 | |
AK201051.1 | - | 3 - 4 | 342 | 94 |