The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008167.2 | NP_032193.1 | 15 - 16 | 2603 | 99 |
GenBank mRNA | AK032669.1 | - | 3 - 4 | 427 | 99 |
BC139823.1 | - | 15 - 16 | 2607 | 99 | |
D13266.1 | - | 15 - 16 | 2603 | 99 |