Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fgd5
Entrez Gene ID
232237
Gene Name
FYVE, RhoGEF and PH domain containing 5
Gene Aliases
C330025N11Rik, ZFYVE23, mFLJ00274
Chromosome Location
Chr.6: 91978821 - 92076005 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172731.2 NP_766319.2 4 - 5 2576 104
GenBank mRNA AK220153.1 - 5 - 6 3050 104
BC060664.1 - 4 - 5 2039 104
BC145502.1 - 5 - 6 2390 104
BC150786.1 - 4 - 5 2525 104

View Full Product Details