Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Adamts9
- Entrez Gene ID
-
101401
- Gene Name
- a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 9
- Gene Aliases
-
1810011L16Rik, 8430403M15Rik, AW743315, E030027K14Rik
- Chromosome Location
-
Chr.6: 92772699 - 92944047 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_175314.3 |
NP_780523.2 |
12 - 13 |
1710 |
71 |
GenBank mRNA |
CK793536.1 |
- |
2 - 3 |
242 |
71 |
View Full Product Details