The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080979.1 | NP_001074448.2 | 9 - 10 | 1456 | 75 |
NM_011567.2 | NP_035697.3 | 10 - 11 | 1585 | 75 | |
GenBank mRNA | AB221623.1 | - | 8 - 9 | 886 | 75 |
AK133915.1 | - | 10 - 11 | 1247 | 75 | |
BC130257.1 | - | 8 - 9 | 891 | 75 | |
D87965.1 | - | 9 - 10 | 1456 | 75 | |
D87966.1 | - | 10 - 11 | 1580 | 75 | |
L26343.1 | - | 10 - 11 | 1125 | 75 | |
L26344.1 | - | - | 1070 | 75 | |
U51743.1 | - | 8 - 9 | 871 | 75 | |
X94441.1 | - | 9 - 10 | 1016 | 75 |