Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crbn
Entrez Gene ID
58799
Gene Name
cereblon
Gene Aliases
2610203G15Rik, 2900045O07Rik, AF229032, AW108261, piL
Chromosome Location
Chr.6: 106778244 - 106800087 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021449.3 NP_067424.2 1 - 2 93 136
GenBank mRNA AK160219.1 - 1 - 2 72 136
BC086488.1 - 1 - 2 71 136

View Full Product Details