The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028099.4 | NP_082375.4 | 8 - 9 | 921 | 90 |
GenBank mRNA | AK008492.1 | - | 3 - 4 | 305 | 90 |
AK081629.1 | - | 8 - 9 | 898 | 90 | |
AK088941.1 | - | 8 - 9 | 918 | 90 | |
BC028640.1 | - | 4 - 5 | 348 | 90 | |
BC067028.1 | - | 8 - 9 | 875 | 90 |