Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rpusd3
Entrez Gene ID
101122
Gene Name
RNA pseudouridylate synthase domain containing 3
Gene Aliases
AI527266
Chromosome Location
Chr.6: 113415319 - 113419459 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033204.1 NP_001028376.1 1 - 2 125 120
GenBank mRNA AK076749.1 - 1 - 2 126 120
AK133871.1 - 1 - 2 105 120
BC116825.1 - 1 - 2 125 120
BC116827.1 - 1 - 2 125 120
BC144733.1 - 1 - 2 125 120
BC144734.1 - 1 - 2 125 120

View Full Product Details