Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Themis
- Entrez Gene ID
-
210757
- Gene Name
- thymocyte selection associated
- Gene Aliases
-
E430004N04Rik, Gasp, Spot, Tsepa, thylex
- Chromosome Location
-
Chr.10: 28668327 - 28883820 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK041386.1 |
- |
4 - 5 |
2061 |
85 |
View Full Product Details