Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rfx6
Entrez Gene ID
320995
Gene Name
regulatory factor X, 6
Gene Aliases
4930572O07Rik, Rfxdc1
Chromosome Location
Chr.10: 51677752 - 51730432 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159389.1 NP_001152861.1 14 - 15 1600 87
NM_177306.3 NP_796280.1 11 - 12 1231 87
GenBank mRNA AK029809.1 - 11 - 12 1232 87
BC132104.1 - 9 - 10 914 87
BC132294.1 - 9 - 10 914 87
FJ804414.1 - 13 - 14 1448 87

View Full Product Details