Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Grm7
- Entrez Gene ID
-
108073
- Gene Name
- glutamate receptor, metabotropic 7
- Gene Aliases
-
6330570A01Rik, BB176677, C030018L03, E130018M02Rik, Gpr1g, Gprc1g, mGluR7
- Chromosome Location
-
Chr.6: 110645458 - 111567230 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK053447.1 |
- |
2 - 3 |
841 |
67 |
View Full Product Details