Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem69
Entrez Gene ID
230657
Gene Name
transmembrane protein 69
Gene Aliases
A630048M13Rik
Chromosome Location
Chr.4: 116551528 - 116557042 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177670.4 NP_808338.2 1 - 2 67 136
GenBank mRNA AK041960.1 - 1 - 2 52 136
BC106178.1 - 1 - 2 60 136

View Full Product Details