Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Man1a
Entrez Gene ID
17155
Gene Name
mannosidase 1, alpha
Gene Aliases
Man1a1, PCR1
Chromosome Location
Chr.10: 53904785 - 54076586 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008548.4 NP_032574.1 10 - 11 2066 76
GenBank mRNA AK031038.1 - 10 - 11 2067 76
AK042371.1 - 9 - 10 1907 76
AK043932.1 - 10 - 11 2007 76
AK154820.1 - 10 - 11 2008 76
AK171841.1 - 10 - 11 2009 76
BC015265.1 - 10 - 11 1967 76
U04299.1 - 9 - 10 1670 76

View Full Product Details