Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Hivep2
- Entrez Gene ID
-
15273
- Gene Name
- human immunodeficiency virus type I enhancer binding protein 2
- Gene Aliases
-
MIBP1, Schnurri-2, Shn-2
- Chromosome Location
-
Chr.10: 13965779 - 14151378 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK040673.1 |
- |
3 - 4 |
231 |
105 |
View Full Product Details