Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myo1g
Entrez Gene ID
246177
Gene Name
myosin IG
Gene Aliases
E430002D17Rik
Chromosome Location
Chr.11: 6506548 - 6520960 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178440.4 NP_848534.2 12 - 13 1694 97
GenBank mRNA AK030825.1 - 12 - 13 1695 97
AK036461.1 - 12 - 13 1679 97
AK042033.1 - 12 - 13 1646 97
AK088011.1 - 12 - 13 1695 97
AK146251.1 - 12 - 13 1667 97
AK152797.1 - 12 - 13 1688 97
AK159700.1 - 12 - 13 1670 97
AK171284.1 - 12 - 13 1688 97
AK171505.1 - 12 - 13 1667 97
BC113755.1 - 12 - 13 1638 97
BC113954.1 - 12 - 13 1638 97

View Full Product Details