Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Esr1
- Entrez Gene ID
-
13982
- Gene Name
- estrogen receptor 1 (alpha)
- Gene Aliases
-
ER, ER-alpha, ERa, ERalpha, ESR, Estr, Estra, Nr3a1
- Chromosome Location
-
Chr.10: 4611989 - 5005633 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK039911.1 |
- |
1 - 2 |
144 |
69 |
View Full Product Details