Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vstm2a
Entrez Gene ID
211739
Gene Name
V-set and transmembrane domain containing 2A
Gene Aliases
Vstm2
Chromosome Location
Chr.11: 16257602 - 16284551 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK079096.1 - - 1088 76

View Full Product Details