Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ankrd36
- Entrez Gene ID
-
76389
- Gene Name
- ankyrin repeat domain 36
- Gene Aliases
-
1700008J08Rik, 1700012M14Rik, Ankrd48
- Chromosome Location
-
Chr.11: 5569680 - 5693796 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK005775.1 |
- |
6 - 7 |
1435 |
118 |
View Full Product Details