Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dbnl
Entrez Gene ID
13169
Gene Name
drebrin-like
Gene Aliases
Abp1, SH3P7, mAbp1
Chromosome Location
Chr.11: 5788483 - 5800980 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013810.3 NP_038838.1 7 - 8 787 76
GenBank mRNA AK140752.1 - 7 - 8 754 76
AK152110.1 - 7 - 8 752 76
AK161973.1 - 7 - 8 76
U58884.1 - 7 - 8 740 76

View Full Product Details