The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178713.4 | NP_848828.1 | 6 - 7 | 1045 | 83 |
GenBank mRNA | AF510322.1 | - | 6 - 7 | 1054 | 83 |
AK050298.1 | - | 6 - 7 | 1046 | 83 | |
AK143752.1 | - | 6 - 7 | 1046 | 83 | |
BC013511.1 | - | 4 - 5 | 587 | 83 | |
BC038493.1 | - | 6 - 7 | 1057 | 83 |