The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136067.2 | NP_001129539.1 | 4 - 5 | 654 | 73 |
NM_001161422.1 | NP_001154894.1 | 3 - 4 | 472 | 73 | |
NM_001161423.1 | NP_001154895.1 | 3 - 4 | 524 | 73 | |
NM_010547.2 | NP_034677.2 | 3 - 4 | 550 | 73 | |
GenBank mRNA | AF069542.1 | - | 2 - 3 | 409 | 73 |
AF513109.1 | - | 4 - 5 | 565 | 73 | |
AK148163.1 | - | 3 - 4 | 508 | 73 | |
AK154095.1 | - | 3 - 4 | 536 | 73 | |
AK188269.1 | - | 1 - 2 | 136 | 73 | |
AY112937.1 | - | 4 - 5 | 654 | 73 | |
CN701499.1 | - | 3 - 4 | 472 | 73 | |
CN717929.1 | - | 3 - 4 | 465 | 73 |