The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016981.2 | NP_058677.1 | 1 - 2 | 1136 | 63 |
GenBank mRNA | AK150135.1 | - | 1 - 2 | 1139 | 63 |
AK154857.1 | - | 1 - 2 | 1173 | 63 | |
AK165501.1 | - | 1 - 2 | 1136 | 63 | |
BC051431.1 | - | 1 - 2 | 483 | 63 | |
BC052708.1 | - | 1 - 2 | 1018 | 63 | |
U51112.1 | - | 1 - 2 | 679 | 63 |