The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122958.1 | NP_001116430.1 | 17 - 18 | 2166 | 83 |
NM_001122959.1 | NP_001116431.1 | 17 - 18 | 2145 | 83 | |
NM_009015.3 | NP_033041.3 | 16 - 17 | 2496 | 83 | |
GenBank mRNA | AK088876.1 | - | 17 - 18 | 2163 | 83 |
AK161644.1 | - | 17 - 18 | 2119 | 83 | |
BC021643.1 | - | 16 - 17 | 2477 | 83 | |
X97796.1 | - | 16 - 17 | 83 |