The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037916.3 | NP_001033005.1 | 11 - 12 | 1514 | 67 |
GenBank mRNA | AK003161.1 | - | 1 - 2 | 105 | 67 |
AK029257.1 | - | 11 - 12 | 1516 | 67 | |
AK076542.1 | - | 10 - 11 | 1485 | 67 | |
BC150874.1 | - | 9 - 10 | 1301 | 67 | |
BC172023.1 | - | 9 - 10 | 1301 | 67 |