Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sobp
Entrez Gene ID
109205
Gene Name
sine oculis-binding protein homolog (Drosophila)
Gene Aliases
2900009C16Rik, 5330439J01Rik, JXC1, jc
Chromosome Location
Chr.10: 43002500 - 43174588 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175407.3 NP_780616.4 1 - 2 548 100
GenBank mRNA BC057055.1 - 1 - 2 227 100
BC059851.1 - 1 - 2 548 100
DQ157775.1 - 1 - 2 107 100

View Full Product Details