Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sec63
Entrez Gene ID
140740
Gene Name
SEC63-like (S. cerevisiae)
Gene Aliases
5730478J10Rik, AI649014, AW319215
Chromosome Location
Chr.10: 42761496 - 42832514 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153055.3 NP_694695.3 14 - 15 1710 85
GenBank mRNA AK036629.1 - 14 - 15 1401 85
AK049737.1 - 12 - 13 3831 85
AY024346.1 - 14 - 15 1507 85
AY238936.1 - 14 - 15 1480 85
BC019366.1 - 14 - 15 1474 85
BC029774.1 - 6 - 7 714 85
BC031846.1 - 14 - 15 1474 85
BC059816.1 - 14 - 15 1709 85
BC080810.1 - 13 - 14 3013 85

View Full Product Details