Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxj3
Entrez Gene ID
230700
Gene Name
forkhead box J3
Gene Aliases
C330039G02Rik, Fhd6
Chromosome Location
Chr.4: 119539661 - 119629119 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290696.1 NP_001277625.1 6 - 7 863 119
NM_172699.3 NP_766287.1 7 - 8 965 119
GenBank mRNA AK082840.1 - 7 - 8 966 119
AK129271.1 - 6 - 7 836 119
AK139508.1 - 7 - 8 899 119
BC058231.1 - 7 - 8 938 119
CB245622.1 - 4 - 5 544 119

View Full Product Details