The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014900.2 | NP_001014900.2 | 1 - 2 | 309 | 61 |
GenBank mRNA | AY959941.1 | - | 1 - 2 | 120 | 61 |
BC147552.1 | - | 1 - 2 | 219 | 61 | |
BC147553.1 | - | 1 - 2 | 219 | 61 | |
BC147848.1 | - | 1 - 2 | 309 | 61 | |
BC147851.1 | - | 1 - 2 | 309 | 61 |