Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fuca1
Entrez Gene ID
71665
Gene Name
fucosidase, alpha-L- 1, tissue
Gene Aliases
0610006A03Rik, 9530055J05Rik, Afuc, Fuca
Chromosome Location
Chr.4: 135920726 - 135940300 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024243.4 NP_077205.3 2 - 3 556 80
GenBank mRNA AK002230.1 - 2 - 3 524 80
AK053323.1 - 2 - 3 501 80
AK089958.1 - 2 - 3 526 80
AK150526.1 - 2 - 3 491 80
AK150890.1 - 2 - 3 548 80
AK151258.1 - 2 - 3 524 80
AK151361.1 - 2 - 3 545 80
AK151879.1 - 2 - 3 545 80
AK151908.1 - 2 - 3 499 80
AK152070.1 - 2 - 3 499 80
AK165604.1 - 2 - 3 526 80
BC003235.1 - 2 - 3 529 80

View Full Product Details