Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcea3
Entrez Gene ID
21401
Gene Name
transcription elongation factor A (SII), 3
Gene Aliases
S-II, SII-K1
Chromosome Location
Chr.4: 136247936 - 136274903 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC010807.1 - 10 - 11 1116 77
D83387.1 - 10 - 11 1065 77

View Full Product Details