Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lao1
Entrez Gene ID
100470
Gene Name
L-amino acid oxidase 1
Gene Aliases
AW990848
Chromosome Location
Chr.4: 118961967 - 118968910 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133892.4 NP_598653.3 5 - 6 661 64
GenBank mRNA AB034801.2 - 5 - 6 661 64
AK052781.1 - 5 - 6 632 64
BC017599.1 - 5 - 6 1050 64
BC023198.1 - 4 - 5 507 64

View Full Product Details