Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rtn4ip1
Entrez Gene ID
170728
Gene Name
reticulon 4 interacting protein 1
Gene Aliases
D10Ertd690e, NIMP
Chromosome Location
Chr.10: 43901807 - 43947862 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130892.4 NP_570962.2 6 - 7 1259 74
GenBank mRNA AF336862.1 - 6 - 7 989 74
AK050324.1 - 6 - 7 1257 74
AK085858.1 - 6 - 7 1239 74
AK088029.1 - 6 - 7 1260 74
BC024116.1 - 6 - 7 1229 74

View Full Product Details