The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009566.4 | NP_033592.2 | 5 - 6 | 423 | 88 |
GenBank mRNA | AK141522.1 | - | 5 - 6 | 424 | 88 |
BC096436.1 | - | 4 - 5 | 474 | 88 | |
U47104.1 | - | 5 - 6 | 443 | 88 |