Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hs3st5
Entrez Gene ID
319415
Gene Name
heparan sulfate (glucosamine) 3-O-sulfotransferase 5
Gene Aliases
D930005L05Rik, Gm1151, Hs3ost5
Chromosome Location
Chr.10: 36506804 - 36834397 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253355.1 NP_001240284.1 2 - 3 869 80
GenBank mRNA AK031910.1 - 2 - 3 859 80
BC147490.1 - 2 - 3 862 80
BC150973.1 - 2 - 3 862 80
BC158004.1 - 2 - 3 862 80

View Full Product Details