Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asb3
Entrez Gene ID
65257
Gene Name
ankyrin repeat and SOCS box-containing 3
Gene Aliases
2400011J03Rik, Asb-3
Chromosome Location
Chr.11: 30954194 - 31102704 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF155354.1 - 8 - 9 1373 61
BC023086.1 - 9 - 10 1492 61

View Full Product Details