The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284359.1 | NP_001271288.1 | 6 - 7 | 753 | 89 |
NM_001284360.1 | NP_001271289.1 | 4 - 5 | 584 | 89 | |
NM_181569.3 | NP_853547.1 | 5 - 6 | 714 | 89 | |
NR_104306.1 | 5 - 6 | 632 | 89 | ||
GenBank mRNA | AK036405.1 | - | 5 - 6 | 623 | 89 |
AK036633.1 | - | 4 - 5 | 683 | 89 | |
AK039600.1 | - | 4 - 5 | 560 | 89 | |
AK047190.1 | - | 6 - 7 | 662 | 89 | |
AK081902.1 | - | 5 - 6 | 625 | 89 | |
AK086378.1 | - | - | 527 | 89 | |
AK090290.1 | - | - | 492 | 89 | |
AK154884.1 | - | 5 - 6 | 584 | 89 | |
BC064471.1 | - | 5 - 6 | 714 | 89 |