The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177973.1 | NP_001171444.1 | 6 - 7 | 972 | 91 |
NM_001177974.1 | NP_001171445.1 | 6 - 7 | 969 | 91 | |
NM_001177975.1 | NP_001171446.1 | 6 - 7 | 969 | 91 | |
NM_001177976.1 | NP_001171447.1 | 6 - 7 | 969 | 91 | |
NM_008363.2 | NP_032389.2 | 6 - 7 | 969 | 91 | |
GenBank mRNA | AB088370.1 | - | 6 - 7 | 796 | 91 |
AF103876.1 | - | 6 - 7 | 969 | 91 | |
AK045958.1 | - | 6 - 7 | 878 | 91 | |
AK143694.1 | - | 6 - 7 | 880 | 91 | |
AK144879.1 | - | 6 - 7 | 878 | 91 | |
AK210148.1 | - | 2 - 3 | 236 | 91 | |
AY184362.1 | - | 6 - 7 | 969 | 91 | |
AY184363.1 | - | 6 - 7 | 969 | 91 | |
AY184364.1 | - | 6 - 7 | 969 | 91 | |
BC004778.1 | - | 6 - 7 | 964 | 91 | |
U56773.2 | - | 6 - 7 | 793 | 91 |