Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pnck
Entrez Gene ID
93843
Gene Name
pregnancy upregulated non-ubiquitously expressed CaM kinase
Gene Aliases
Bstk3, CaMKIbeta2, CaMKlb2, Camk1b, Punc, caMKlb1
Chromosome Location
Chr.X: 73655992 - 73660127 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199351.1 NP_001186280.1 6 - 7 732 93
NM_001199352.1 NP_001186281.1 6 - 7 596 93
NM_012040.3 NP_036170.1 6 - 7 672 93
GenBank mRNA AB023027.1 - 5 - 6 542 93
AF181984.1 - 6 - 7 646 93
AK134665.1 - 6 - 7 603 93
AK158755.1 - 6 - 7 597 93
BC051996.1 - 6 - 7 579 93
BC055891.1 - 6 - 7 572 93

View Full Product Details