The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009279.3 | NP_033305.1 | 1 - 2 | 168 | 88 |
GenBank mRNA | AK013723.1 | - | 1 - 2 | 130 | 88 |
AK160089.1 | - | 1 - 2 | 134 | 88 | |
CJ144362.1 | - | 1 - 2 | 173 | 88 | |
X90582.1 | - | 1 - 2 | 249 | 88 |