The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008229.2 | NP_032255.2 | 9 - 10 | 1189 | 86 |
GenBank mRNA | AB221663.1 | - | 9 - 10 | 981 | 86 |
AK135572.1 | - | 4 - 5 | 495 | 86 | |
BC138517.1 | - | 9 - 10 | 1161 | 86 | |
U31758.1 | - | 9 - 10 | 1149 | 86 |