Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gje1
Entrez Gene ID
76743
Gene Name
gap junction protein, epsilon 1
Gene Aliases
AEY12, Cx23, D230044M03Rik, Gjf1, Gsfaey12
Chromosome Location
Chr.10: 14715626 - 14718214 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029722.1 NP_083998.1 2 - 3 291 64
GenBank mRNA AK018698.1 - 2 - 3 292 64
DQ149609.1 - 1 - 2 84 64

View Full Product Details