Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sf3a1
Entrez Gene ID
67465
Gene Name
splicing factor 3a, subunit 1
Gene Aliases
1200014H24Rik, 5930416L09Rik, AI159724
Chromosome Location
Chr.11: 4160354 - 4182541 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026175.5 NP_080451.4 4 - 5 775 97
GenBank mRNA AK004763.1 - 4 - 5 758 97
AK028829.1 - 4 - 5 727 97
AK029095.1 - 4 - 5 745 97
AK030223.1 - 4 - 5 762 97
AK051703.1 - 4 - 5 775 97
AK134178.1 - 4 - 5 754 97
AK160061.1 - 4 - 5 758 97
AK191762.1 - 1 - 2 299 97
AK206768.1 - 1 - 2 275 97
AK215839.1 - 1 - 2 130 97
BC029753.1 - 4 - 5 739 97

View Full Product Details