The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198248.1 | NP_937891.1 | 13 - 14 | 2818 | 71 |
GenBank mRNA | AK220256.1 | - | 14 - 15 | 2703 | 71 |
BC059177.1 | - | 13 - 14 | 2818 | 71 |