Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb40
Entrez Gene ID
230848
Gene Name
zinc finger and BTB domain containing 40
Gene Aliases
BC059177, C230087D24, Gm571, mKIAA0478
Chromosome Location
Chr.4: 136979732 - 137049442 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198248.1 NP_937891.1 17 - 18 3453 61
GenBank mRNA AK048974.1 - 2 - 3 109 61
AK220256.1 - 18 - 19 3338 61
BC059177.1 - 17 - 18 3453 61

View Full Product Details