Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Fam83e
- Entrez Gene ID
-
73813
- Gene Name
- family with sequence similarity 83, member E
- Gene Aliases
-
4930403C10Rik
- Chromosome Location
-
Chr.7: 45721220 - 45729492 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK015058.1 |
- |
1 - 2 |
57 |
57 |
View Full Product Details